VWR UniTranz-RT® Transport System, Media Only
Universal transport media, Liquide, 3 ml
Catalog # 77200-790
Supplier: VWR International
VWR UniTranz-RT® Transport System, Media Only
Catalog # 77200-790
Supplier: VWR International
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The UniTranz-RT® universal transport is ideal for the collection and preservation of viruses including, chlamydia, mycoplasma, and urea plasma. It is also suitable for use with antigen detection, PCR, and rapid tests when validated by test manufacturers and/or laboratory.
- 3 ml fill
- Medium is stable at room temperature
- No swab, media only
- Premium medical grade plastic components improve product shelf life
- Shelf life: 18 months
- For In Vitro diagnostic use
- Made in USA
Contains antimicrobial agents to minimize bacterial and fungal contamination, as well as glass beads to release and disperse sample into the medium during vortexing.
Specifications
Specifications
- Format du milieu:Liquide
- Désignation:Universal transport media
- Taille:3 ml