Cage-Klenz® 180 Alkaline Detergents
Supplier: Steris
New Product
Certificates
About this item
Cage-Klenz® 180 Detergent is a liquid, phosphate-free detergent for use in high pressure spray animal cage washing equipment. It is formulated with corrosion inhibitors for enhanced materials compatibility.
- Efficacy - Effectively cleans animal fats, oils and other organic cage residues in both soft and hard water
- Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
- Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
- Environmentally Friendly - phosphate-free formulation
- Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Cage-Klenz® 180 Detergent is specially formulated to meet the unique cleaning and materials compatibility requirements found in research animal caging.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together