Cage-Klenz® 280 Acid-Based Detergents
Catalogue # 77531-220
Fournisseur: Steris
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Size:19 L (5 gal.)
- Cat. no.:77531-220
- Packaging:Plastic pail
Spécifications
À propos de cet article
Cage-Klenz® 280 Detergent is a phosphoric acid-based detergent for use in high pressure spray animal cage washing equipment.
- Efficacy - Effectively cleans hard water and urine scale, and metal oxides in both soft and hard water
- Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
- Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
- Environmentally Friendly - formulation with biodegradable surfactants
- Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Cage-Klenz® 280 Detergent is an economical detergent specially formulated to remove challenging hard water and urine scales found in research animal caging.