Vee Gee Erlenmeyer Flasks | Avantor
Vee Gee Erlenmeyer Flasks
Supplier:   Heathrow Scientific
Vee Gee Erlenmeyer Flasks
Certificates
Vee Gee Erlenmeyer Flasks
Vee Gee Erlenmeyer Flasks
5 Options Available Below
Product Details & Documents
Erlenmeyer flasks are ideal for a wide variety of applications including cell culture, chemical reactions and microbial cultivation.
  • Made from premium borosilicate glass with a low coefficient of expansion
  • Uniform wall thickness (ideal for heating applications)
  • Graduation accuracy is ±5% of full capacity
  • Heavy-duty rims to reduce chipping
  • Contains permanent white graduations and a large marking square
  • Wide range of sizes to choose from (100 to 2000 ml)
Each flask is constructed out of premium borosilicate glass with a low coefficient of expansion. They also feature heavy-duty rims in order to reduce chipping and have uniform wall thickness to withstand various heating applications.
Product Family Options
Informations sur le produit
  • Capacité
    Intervalle de graduation
    Stopper No.
    Availability
    Price
  • 100 ml
    10 ml
    2
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $88.70
    $88.70
    Prix promotionnel pour les nouveaux clients
    Pack of 10
       $88.70
    Supplier Number: 10530-100A
  • 300 ml
    25 ml
    6
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $108.68
    $108.68
    Prix promotionnel pour les nouveaux clients
    Pack of 10
       $108.68
    Supplier Number: 10530-300A
  • 500 ml
    50 ml
    6
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $184.35
    $184.35
    Prix promotionnel pour les nouveaux clients
    Pack of 10
       $184.35
    Supplier Number: 10530-500A
  • 1000 ml
    50 ml
    8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $227.95
    $227.95
    Prix promotionnel pour les nouveaux clients
    Pack of 10
       $227.95
    Supplier Number: 10530-1000A
  • 200 ml
    200 ml
    10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $75.81
    $75.81
    Prix promotionnel pour les nouveaux clients
    Each
       $75.81
    Supplier Number: 10530-2000