Vee Gee Separatory Funnels
Supplier:   Heathrow Scientific
Vee Gee Separatory Funnels
Certificates
Vee Gee Separatory Funnels
Vee Gee Separatory Funnels
4 Options Available Below
Product Details & Documents
Our separatory funnels are an excellent tool for liquid-liquid extractions, phase separations, and chemical processes in laboratories.
  • Supplied with PTFE stopcocks to eliminate freezing and lubricant contamination
  • Made from high-quality borosilicate tubing with a low coefficient of expansion
  • Features smooth tooled tips, white block letters, and large marking squares
  • Squibb style
Whether you're in a research laboratory, educational institution, or industrial setting, our separatory funnels empower you to achieve the highest standards of accuracy in your separation procedures.
These separatory funnels feature smooth tooled tips, large white block letters, and large marking squares. Each funnel is made from borosilicate tubing with a low coefficient of expansion, and are supplied with PTFE stopcocks, which eliminates freezing and lubricant contamination.
Product Family Options
Product Information
  • Capacité
    Plug Size
    ST Stopper No.
    Availability
    Price
  • 300 ml
    4 (stopcock)
    24/40
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $194.40
    $194.40
    New Customer Promotional Price
    Log in for availability
    Pack of 6
    Pack of 6 - $1,166.38
    $1,166.38
    New Customer Promotional Price
    Each
       $194.40
    Pack of 6
       $1,166.38
    Supplier Number: 1426T-300
  • 500 ml
    4 (stopcock)
    24/40
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $235.19
    $235.19
    New Customer Promotional Price
    Log in for availability
    Pack of 6
    Pack of 6 - $1,411.13
    $1,411.13
    New Customer Promotional Price
    Each
       $235.19
    Pack of 6
       $1,411.13
    Supplier Number: 1426T-500
  • 1000 ml
    6 (stopcock)
    29/42
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $347.78
    $347.78
    New Customer Promotional Price
    Log in for availability
    Pack of 4
    Pack of 4 - $1,391.11
    $1,391.11
    New Customer Promotional Price
    Each
       $347.78
    Pack of 4
       $1,391.11
    Supplier Number: 1426T-1000
  • 2000 ml
    6 (stopcock)
    29/42
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $424.55
    $424.55
    New Customer Promotional Price
    Log in for availability
    Pack of 2
    Pack of 2 - $849.10
    $849.10
    New Customer Promotional Price
    Each
       $424.55
    Pack of 2
       $849.10
    Supplier Number: 1426T-2000