Vee Gee Filtering Flasks
Supplier: Heathrow Scientific
Jump to:
À propos de cet article
Filtering flasks are constructed of heavy wall borosilicate glass in order to withstand high vacuum applications.
- Made of premium borosilicate glass
- Heavy-duty rims to reduce chipping
- Permanent white graduations
- Large marking square for labeling
- Port for tubulation has an outside diameter of 10 mm
- Accepts a No. 8 rubber stopper (Not included)
Used in conjunction with vacuum filter systems, these filtering flasks feature heavy-duty rims to reduce chipping. The flasks also feature permanent white graduations and a large marking square for labeling. All flasks are sold individually.
4 Options Available Below
Product Details & Documents
Filtering flasks are constructed of heavy wall borosilicate glass in order to withstand high vacuum applications.
- Made of premium borosilicate glass
- Heavy-duty rims to reduce chipping
- Permanent white graduations
- Large marking square for labeling
- Port for tubulation has an outside diameter of 10 mm
- Accepts a No. 8 rubber stopper (Not included)
Used in conjunction with vacuum filter systems, these filtering flasks feature heavy-duty rims to reduce chipping. The flasks also feature permanent white graduations and a large marking square for labeling. All flasks are sold individually.
Product Family Options
Informations sur le produit
- CapacitéMatièreO.D.×HAvailabilityPrice
- 300 mlBorosilicate glass3.4×5.5"Log in pour la disponibilitéPack of 12Pack of 12 - $619.13$619.13Prix promotionnel pour les nouveaux clientsEach$51.59Pack of 12$619.13Supplier Number: 1780T-300
- 500 mlBorosilicate glass4.1×6.7"Log in pour la disponibilitéPack of 12Pack of 12 - $712.41$712.41Prix promotionnel pour les nouveaux clientsEach$59.37Pack of 12$712.41Supplier Number: 1780T-500
- 1000 mlBorosilicate glass5.2×8.7"Log in pour la disponibilitéPack of 8Pack of 8 - $639.14$639.14Prix promotionnel pour les nouveaux clientsEach$79.89Pack of 8$639.14Supplier Number: 1780T-1000
- 2000 mlBorosilicate glass6.5×10.6"Log in pour la disponibilitéPack of 4Pack of 4 - $495.66$495.66Prix promotionnel pour les nouveaux clientsEach$123.91Pack of 4$495.66Supplier Number: 1780T-2000