Vee Gee Filter Holder Systems | Avantor
Vee Gee Filter Holder Systems
Supplier:   Heathrow Scientific
Vee Gee Filter Holder Systems
Certificates
Vee Gee Filter Holder Systems
Vee Gee Filter Holder Systems
4 Options Available Below
Product Details & Documents
Filtration filter holders are offered with a wide variety of components for different applications.
  • Available in 100 or 300 ml capacities
Available with a fritted glass support base for general and biological applications, or with a removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
Product Family Options
Informations sur le produit
  • Désignation
    For Use With
    Funnel Capacity
    Availability
    Price
  • Fritted glass support base
    General and biological applications
    100 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $263.47
    $263.47
    Prix promotionnel pour les nouveaux clients
    Each
       $263.47
    Supplier Number: 6168-0003
  • Stainless steel support base
    Ultraclean filtrate and proteinaceous solutions
    100 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $305.94
    $305.94
    Prix promotionnel pour les nouveaux clients
    Each
       $305.94
    Supplier Number: 6168-0004
  • Fritted glass support base
    General and biological applications
    300 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $267.01
    $267.01
    Prix promotionnel pour les nouveaux clients
    Each
       $267.01
    Supplier Number: 6168-0001
  • Stainless steel support base
    Ultraclean filtrate and proteinaceous solutions
    300 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $345.16
    $345.16
    Prix promotionnel pour les nouveaux clients
    Each
       $345.16
    Supplier Number: 6168-0002