Vee Gee Filter Holder Components
Supplier: Heathrow Scientific
Jump to:
À propos de cet article
Available with fritted glass support base for general and biological applications, or with removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
- For use with Vee Gee Filter system kits
All items except the PTFE gasket are autoclavable.
8 Options Available Below
Product Details & Documents
Available with fritted glass support base for general and biological applications, or with removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
- For use with Vee Gee Filter system kits
All items except the PTFE gasket are autoclavable.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 6168-47100
- Supplier Number: 6168-4711
- Supplier Number: 6168-4721
- Supplier Number: 6168-4722
- Supplier Number: 6168-4731
- PTFE gasketsLog in pour la disponibilitéLog in pour la disponibilitéPack of 10Pack of 10 - $18.43$18.43Prix promotionnel pour les nouveaux clientsPack of 10$18.43Supplier Number: 6168-4741
- Supplier Number: 6168-4751
- Supplier Number: 6168-4762