Consistometer, Bostwick
Consistometer
Catalog # 23270-004
Small Business Enterprise
Supplier: C S C Scientific
Supplier #: 24925-000
Jump to:
Product Details & Documents
Determines consistency of almost any viscous substance by measuring the distance a material flows under its own weight during a given time interval
Can be used by food, paint, chemical, and other industries to pre-determine formulas for standardizing production lots. Made of stainless steel, unit has spirit level and two leveling screws. Trigger-held gate is spring-operated to permit instantaneous release. Trough is graduated in 0.5cm divisions. Requires 75mL sample.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Specifications
- Désignation:Consistometer
- Cat. No.:23270-004
- L×W×H:35.6 x 8.9 x 14 cm (14 x 31/2 x 51/2")