CP-811 Emulsifying Detergent/Additive, CANI
Fournisseur: Cani
Certificats
À propos de cet article
Phosphate-free, neutral pH liquid detergent and additive effectively removes a wide range of residues and is particularly effective against oils, creams, grease, petrolatums, and waxes
When used alone or with compatible alkaline cleaners, the detergent’s powerful emulsifiers and enhanced low-foam surfactant blend improve cleaning and rinsing, making it ideal for recirculating spray washers, pharmaceutical/biotech process cleaning, cosmetic process cleaning, and laboratory glassware washing. Detergent rinses completely for a clean, residue-free surface.
Frequently Bought Together