ContactUsLinkComponent
Surgical Instrument Stain Remover, Integra™ Miltex®
Surgical Instrument Stain Remover, Integra™ Miltex®
Catalog # 95042-716
Supplier:  Miltex
undefined
Surgical Instrument Stain Remover, Integra™ Miltex®
Catalog # 95042-716
Supplier:  Miltex
Supplier Number:  3-740
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Jar
  • Size:
    85 g (3 oz.)
  • Cat. no.:
    95042-716
  • Supplier No.:
    3-740
Specifications
About this item
Powdered formula removes stains and tarnish, and restores luster to surgical instruments, autoclaves, ultrasonic cleaners, stainless steel countertops, tiles, and plated surfaces
Effective on a variety of materials including stainless steel, chrome, nickel, brass, copper, and silver. The formula will not harm instrument surfaces.
Ordering information: Stain remover is packaged in an 85g (3oz.) jar. Jars are supplied in cases of 12.