neodisher® Detergents, Miele
neodisher® Detergents, Miele
Supplier:   Miele
neodisher® Detergents, Miele
Supplier:  Miele
Certificates
neodisher® Detergents, Miele
neodisher® Detergents, Miele
2 Options Available Below
Product Details & Documents
These powder and liquid detergents provide consistent, high level cleanliness in Miele glassware washing systems
Detergents FA and MA are for the removal of creams and lotions, cleaning stainless steel, and washing dental instruments. Detergents FT and F are for the removal of inks. Detergent FT, as well as GK, is also used for cell and tissue culture residues. Detergent FLA works well with wastewater, food, environmental, educational, and chemical applications. Detergent A8 is suited for generalized cleaning; detergent LA removes fats, oils, and waxes in food applications; and detergent UW is suitable for trace metal removal from glassware. Neutralizing agent N is composed of phosphoric and organic acids, while agent Z is composed only of organic acids. Both are meant to be dispensed into the rinse water after the use of alkaline-based detergents.
Ideal for the removal of organic, inorganic, and metal residues from laboratory glassware and medical and dental instruments in many industries, including cosmetic, wastewater, environmental, biotech, and food.
Documents
Product Family Options
Product Information
  • Désignation
    Taille
    Availability
    Price
  • Neodisher EM 5L
    5 L
    Log in for availability
    Log in for availability
    Each
    Each - $486.00
    $486.00
    Each
       $486.00
    Minimum order quantity is 1
    Supplier Number: 94000140
  • Neodisher Medizym
    5 L
    Log in for availability
    Log in for availability
    Each
    Each - $267.00
    $267.00
    Each
       $267.00
    Minimum order quantity is 1
    Supplier Number: 94069013