A non-flammable, heavy-duty cleaner and degreaser that removes grease, adhesives, and photo-resistant stains from all general surfaces. Filtered to 0.1 microns, this cleaner is ideal for semiconductor, electronics, aerospace, and clean-build operations.
- Non-flammable, heavy-duty cleanroom degreaser
- Effective at low concentrations
- Filtered to 0.1 µm
- Biodegradable
- Water soluble
The Micronova MegaClean™ heavy-duty cleaner offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Available in 1 or 5 gallon bottles.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 68400-044
- 68400-042
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health