ContactUsLinkComponent
EZ-Zyme® All-Purpose Enzyme Cleaner, Integra™ Miltex®
EZ-Zyme® All-Purpose Enzyme Cleaner, Integra™ Miltex®
Catalog # 21909-148
Supplier:  Miltex
undefined
EZ-Zyme® All-Purpose Enzyme Cleaner, Integra™ Miltex®
Catalog # 21909-148
Supplier:  Miltex
Supplier Number:  3-750
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Dispenser Box contains 32, size 22mL packets.
  • Size:
    22 mL (0.74 oz.)
  • Cat. no.:
    21909-148
  • Supplier No.:
    3-750
Specifications
About this item
Easy-to-use, neutral pH, multi-use cleanser can be used as an instrument presoak, ultrasonic cleaner, laundry presoak, or evacuation system cleaner
It is ideal for cleaning proteinaceous (mucus and tissue) and lipid (fatty) materials from surgical or dental instruments, linen, or water submersible equipment. An excellent alternative to mechanical scrubbing.
Ordering information: Dispenser box contains thirty-two 22mL (3/4oz.) packets (21909-148) that will produce 32 gallons of solution. The 1 gallon bottle (95042-730) will produce 170 gallons of solution.