LabSolutions™ Detergents, Labconco® | Avantor
LabSolutions™ Detergents, Labconco®
LabSolutions™ Detergents, Labconco®
Supplier:  Labconco
Certificates
LabSolutions™ Detergents, Labconco®
LabSolutions™ Detergents, Labconco®
4 Options Available Below
Product Details & Documents
Formulated for use in automatic laboratory glassware washers
Low-Foaming Liquid Detergent removes a wide range of laboratory contaminants. Noncorrosive formula is safe for use on stainless steel, glass, or plastic. Phosphate-free for environmentally safe disposal and chlorine-free for safe use tissue cultures. Can be used in hot or cold water. Comes in a 3.8L (1gal.) jug. Washes approximately 180 loads.
Non-Foaming Powder Detergent effectively removes grease, blood, agar, and protein digestates from glassware. Special formula prevents damage to machine pumps, seals, and gaskets. A 1% solution has an approximate pH of 12.5. Available in a 4.5kg (10lbs.) or 12.5kg (27.5lbs.) bucket.
Neutralizing Acid Rinse removes hard water deposits, such as mineral deposits, accumulated salts, and lime scale. Mild acid corrects pH, reducing alkaline detergent residue. Comes in a 1L (32oz.) bottle. Fills reservoir approximately six times.
Specially designed for the Labconco® SteamScrubber® and FlaskScrubber® washers. Available in liquid, powder, and a neutralizing acid rinse. All formulations rinse clean away.
Documents
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • LabSolutions™ Non-Foaming Powder Detergent, bucket
    4.5 kg (10 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (10lb)
    Each (10lb) - $268.00
    $268.00
    Each (10lb)
       $268.00
    La quantité minimale de commande est de 1
    Supplier Number: 4422000
  • LabSolutions™ Non-Foaming Powder Detergent, bucket
    12.5 kg (27.5 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (440oz)
    Each (440oz) - $539.00
    $539.00
    Each (440oz)
       $539.00
    La quantité minimale de commande est de 1
    Supplier Number: 4422100
  • LabSolutions™ Low-Foaming Liquid Detergent
    3.8 L (1 gal.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1US Gal)
    Each (1US Gal) - $262.00
    $262.00
    Each (1US Gal)
       $262.00
    La quantité minimale de commande est de 1
    Supplier Number: 4522000
  • LabSolutions™ Neutralizing Acid Rinse
    1 L (33.8 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (34oz)
    Each (34oz) - $52.30
    $52.30
    Each (34oz)
       $52.30
    La quantité minimale de commande est de 1
    Supplier Number: 4522200