Anti-Static Wire, Justrite® | Avantor
Anti-Static Wire, Justrite®
Antistatic insulated wire
Catalog # 56610-593
Supplier:   Justrite
Fournisseur #:  08505
Anti-Static Wire, Justrite®
Catalog # 56610-593
Supplier:  Justrite
Anti-Static Wire, Justrite®
Anti-Static Wire, Justrite®
Antistatic insulated wire
Price
114,08 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Prevents sparks from static electricity generated by movement and flow of flammable liquids.
Bonding between containers during liquid transfer and connecting drums to an earth ground are required under several Federal and Local Codes. Ensure adherence to these codes with this flexible 1 m (3') long insulated wire with 1.5 cm (5/8") C-clamp and 1.5 cm (5/8") alligator clip.
Spécifications
  • Désignation:
    Antistatic insulated wire