Drum Opener, Wheaton
Drum Opener
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 885295
Jump to:
Product Details & Documents
Drum opener features a quarter-turn opening action and is cast from a light-weight, durable, sparkless aluminum alloy.
- For use with hazardous material
Documents
Certificates
Recherche par numéro de lotSpécifications
- Désignation:Drum Opener