Precleaned VOA Vials, Environmental Express®
Precleaned VOA Vials, Environmental Express®
Precleaned VOA Vials, Environmental Express®
Certificates
Precleaned VOA Vials, Environmental Express®
Precleaned VOA Vials, Environmental Express®
19 Options Available Below
Product Details & Documents
These volatile organic analysis (VOA) vials are made of borosilicate glass. Choose from an open-top cap (0.125" thick PTFE/silicone septa) or closed-top style cap in a variarty of capacities and clear or amber glass.
Preserved vials come pre-filled with common preservative reagents for convenience and safety.
Certifications: All containers meet one of two levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level I indicates that each case of containers has been custody-sealed, lot number-labeled for traceability, and a Certificate of Analysis indicating that the containers received full EPA quality assurance treatment.
Accessories information: The accessory Foam Block Rack holds 72×40 ml vials. Ideal for storage and shipping 40 ml VOA vials.
Documents
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Color
    Avec bouchon
    Cap Type
    Availability
    Price
  • Level I quality assured vials
    20.00 ml
    Clear
    Oui
    Closed top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $606.12
    $606.12
    Case of 72
       $606.12
    Supplier Number: APC1675P
  • Level I quality assured vials
    60.00 ml
    Clear
    Oui
    Closed top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 144
    Case of 144 - $442.64
    $442.64
    Case of 144
       $442.64
    Supplier Number: APC1559
  • Level I quality assured vials
    40.00 ml
    Amber
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $800.20
    $800.20
    Case of 72
       $800.20
    Supplier Number: APC1590P
  • Level I quality assured vials
    40.00 ml
    Amber
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 144
    Case of 144 - $405.52
    $405.52
    Case of 144
       $405.52
    Supplier Number: APC1541
  • Level I quality assured vials
    40.00 ml
    Clear
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 144
    Case of 144 - $357.45
    $357.45
    Case of 144
       $357.45
    Supplier Number: APC1545
  • Level I quality assured vials
    60.00 ml
    Clear
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $240.81
    $240.81
    Prix promotionnel pour les nouveaux clients
    Case of 72
       $240.81
    Supplier Number: APC1553P
  • Level I quality assured vials, Method 5035
    40.00 ml
    Clear
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $632.33
    $632.33
    Case of 72
       $632.33
    Supplier Number: APC1560
  • Level I quality assured/preserved vials with 0.2 ml Sulfuric Acid and 0.06" Thick PTFE Silicone Bonded Septa
    40.00 ml
    Amber
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $676.61
    $676.61
    Case of 72
       $676.61
    Supplier Number: APC1624
  • Level I quality assured/preserved vials with 1:1 HCL 0.2 ml
    40.00 ml
    Clear
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $668.04
    $668.04
    Case of 72
       $668.04
    Supplier Number: APC1601
  • Level I quality assured/preserved vials with 1:1 HCL 0.5 ml, Method 5035
    40.00 ml
    Amber
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $845.77
    $845.77
    Case of 72
       $845.77
    Supplier Number: APC1628
  • Level I quality assured/preserved vials with 1:1 HCL 1 ml
    40.00 ml
    Clear
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $212.64
    $212.64
    Case of 72
       $212.64
    Supplier Number: APC1598
  • Level I quality assured/preserved vials with 2% Sodium Thiosulfate 0.2 ml
    40.00 ml
    Clear
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $262.87
    $262.87
    Case of 72
       $262.87
    Supplier Number: APC1556
  • Level I quality assured/preserved vials with 5 ml Sodium Bisulfate, stir bar, low level tare weight, Method 5035
    40.00 ml
    Clear
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $1,131.51
    $1,131.51
    Case of 72
       $1,131.51
    Supplier Number: APC1618
  • Level I quality assured/preserved vials with DI Water 5 ml
    40.00 ml
    Clear
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $508.08
    $508.08
    Case of 72
       $508.08
    Supplier Number: APC1630
  • Level I quality assured/preserved vials with Methanol 5 ml
    40.00 ml
    Clear
    Oui
    Open top
    Discontinued
    Supplier Number: APC1596
  • Level III vials
    20.00 ml
    Clear
    Oui
    Closed top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $440.03
    $440.03
    Case of 72
       $440.03
    Supplier Number: APC3675P
  • Level III vials
    40.00 ml
    Clear
    Oui
    Closed top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $271.71
    $271.71
    Case of 72
       $271.71
    Supplier Number: APC3660P
  • Level III vials
    40.00 ml
    Amber
    Oui
    Closed top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $480.45
    $480.45
    Case of 72
       $480.45
    Supplier Number: APC3650
  • Level III vials
    40.00 ml
    Clear
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $374.40
    $374.40
    Case of 72
       $374.40
    Supplier Number: APC3550P
  • Level III vials
    40.00 ml
    Amber
    Oui
    Open top
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $575.39
    $575.39
    Case of 72
       $575.39
    Supplier Number: APC3590P