VWR® Porous-Bottom Crucibles, Porcelain
Fournisseur: VWR International
Certificats
À propos de cet article
Chemical-porcelain crucibles feature a porous bottom with a porous disc permanently formed into the walls.
- Stable porosity
- Consistent rate of flow
- Chemical-porcelain construction
- Cover is ordered separately.
Frequently Bought Together