Staining Dish, 50 Slide Unit, Wheaton
50-Slide Staining Dish with Cover and Rack
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 900400
Jump to:
Product Details & Documents
The 50-Slide Unit Staining Dish accommodates up to 50 microscope slides
Slide sizes acceptable for use are 3x1" (75x25mm), 3x11/2" (75x38mm), and 3x2" (75x51mm). The rack is made of non-tarnsishable stainless steel, and is resistant to staining solutions. The handle is permanently attached, but hinged to permit the closure of the dish and easy insertion and removal of microscope slides. The dish and cover are manufactured from soda-lime glass, with approximate inside bottom dimensions of 185 Lx88Wx78Hmm.
Spécifications
- Désignation:50-Slide Staining Dish with Cover and Rack
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- 50 Slide Staining Dish Only
Informations sur le produit
- Catalog #AvailabilityPrice
- 50 Slide Staining Dish Cover Only
Informations sur le produit
- Catalog #AvailabilityPrice
- 50 Slide Stainless Steel Rack, with Attached Handle50 Slide Stainless Steel Rack, with Attached Handle25461-032Log in pour la disponibilitéCase of 3 - $415.87
Informations sur le produit
- Catalog #AvailabilityPrice
- 50 Slide Staining Dish with Cover, Soda-Lime glass50 Slide Staining Dish with Cover, Soda-Lime glass25461-030Log in pour la disponibilitéCase of 3 - $1,064.70