Synthware Conical Reaction Vials, Heavy Wall
Fournisseur: Kemtech America
Certificats
À propos de cet article
Graduated.[ST] Joint No.: 14/10.
- Heavy wall Borosilicate construction
- Threaded Ground glass joint
- Graduated
- Cap included
This heavy wall reaction vial is great for microscale reactions.
Ordering information: For replacement caps, Viton® O-rings, or septa, contact your Avantor representative.
Delivery: Supplied with one compression cap, one Viton® O-ring, and one fluoropolymer resin-faced silicone rubber septum.
Frequently Bought Together