Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Somso® Lancelet

Somso® Lancelet

Lancelet cross section, 12 cm, 12 cm, 20 cm
Catalog # 470312-482
Supplier:   MARCUS SOMMER
Supplier #:  ZOS59/N
Somso® Lancelet
Catalog # 470312-482
Supplier:  MARCUS SOMMER
Somso® Lancelet
Somso® Lancelet

Lancelet cross section, 12 cm, 12 cm, 20 cm

Price
$951.00
Log in for availability
Each
Product Details & Documents

Somso® premium quality models.

  • Five year warranty

Cross section through the branchia and middle intestine regions, enlarged approx. 150 times, in SOMSO-Plast®. In one piece. On a stand with green base.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.

Specifications
  • Désignation:
    Lancelet cross section
  • Hauteur:
    20 cm
  • Longueur:
    12 cm
  • Width:
    12 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.