Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Somso® Artificial Human Skeleton Fm | Avantor

Somso® Artificial Human Skeleton Fm

Artificial human skeleton fm, 55 cm, 55 cm, 180 cm
Catalog # 470222-856
Supplier:   MARCUS SOMMER
Fournisseur #:  QS 10/7
Somso® Artificial Human Skeleton Fm
Catalog # 470222-856
Supplier:  MARCUS SOMMER
Somso® Artificial Human Skeleton Fm
Somso® Artificial Human Skeleton Fm

Artificial human skeleton fm, 55 cm, 55 cm, 180 cm

Price
$3,340.00
Log in pour la disponibilité
Each
Product Details & Documents

Somso® premium quality models.

  • Five year warranty

Natural cast of the bones of a female adult, in SOMSO-Plast®. Life-like representation of bone structure with full anatomical detail. Skull with removable vault and mandible. Joints movable, upper and lower extremities can be removed. Left and right feet can be removed. Mounted on a stand.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.

Spécifications
  • Désignation:
    Artificial human skeleton fm
  • Hauteur:
    180 cm
  • Longueur:
    55 cm
  • Width:
    55 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.