Polypropylene Vials, Starplex®
Supplier: Starplex
Jump to:
À propos de cet article
These vials are made of crack-resistant medical grade polypropylene.
- With or without screw caps
- Flat, conical or round bottoms
Available clear for easy visual inspection and amber for light-sensitive components. These vials provide convenient storage for samples.
7 Options Available Below
Product Details & Documents
These vials are made of crack-resistant medical grade polypropylene.
- With or without screw caps
- Flat, conical or round bottoms
Available clear for easy visual inspection and amber for light-sensitive components. These vials provide convenient storage for samples.
Product Family Options
Informations sur le produit
- DésignationCapacitéBottom StyleColorAvec bouchonCap TypeCap ColorSterilityAvailabilityPrice
- Unlabeled10.00 mlRound bottomAmberNonNone—Non sterileLog in pour la disponibilitéCase of 1,500$690.00Supplier Number: V8095
- Unlabeled10.00 mlConical bottomClearOuiScrew cap, packed separatelyYellowNon sterileLog in pour la disponibilitéCase of 1,000$267.00Supplier Number: V112-YNL
- Unlabeled10.00 mlFlat bottomClearOuiScrew cap, packed separatelyWhiteNon sterileLog in pour la disponibilitéCase of 1,000$390.00Supplier Number: V102-NL
- Labeled10.00 mlFlat bottomClearOuiScrew cap, packed separatelyWhiteNon sterileLog in pour la disponibilitéCase of 1,000$403.00Supplier Number: V102
- Unlabeled10.00 mlFlat bottomAmberOuiScrew cap, packed separatelyWhiteNon sterileLog in pour la disponibilitéCase of 1,000$397.00Supplier Number: V103-NL
- Labeled10.00 mlFlat bottomClearOuiScrew cap, attached, in plastic peel packWhiteSterileLog in pour la disponibilitéCase of 100$350.00Supplier Number: V102-POUCH
- Unlabeled10.00 mlConical bottomClearNonNone—Non sterileLog in pour la disponibilitéCase of 1,500 (10ml)$296.00Supplier Number: V8094
- Unlabeled10.00 mlConical bottomClearNonNone—SterileDiscontinued-Supplier Number: V8084-S