Screw-Thread Sample Storage Vials, Thermo Scientific | Avantor
Screw-Thread Sample Storage Vials, Thermo Scientific
Screw-Thread Sample Storage Vials, Thermo Scientific
Supplier:  Thermo Scientific
Certificates
Screw-Thread Sample Storage Vials, Thermo Scientific
Screw-Thread Sample Storage Vials, Thermo Scientific
2 Options Available Below
Product Details & Documents
Manufactured under the most stringent conditions, these vials eliminate leaching of ions and provide a consistent pH for the duration of the sample storage.
Available in clear and amber borosilicate glass. Clear glass is Type I, Class A, 33 expansion borosilicate glass. Choose amber glass for light-sensitive samples.
Ordering information: Caps for these vials are sold separately.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Diameter ×H
    Color
    Avec bouchon
    Taille du bouchon
    Availability
    Price
  • Vials
    2.00 ml
    12×32 mm
    Clear
    Non
    8-425
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $127.11
    $127.11
    Prix promotionnel pour les nouveaux clients
    Pack of 100
       $127.11
    Supplier Number: B7999-1
  • Vials
    4.00 ml
    15×45 mm
    Clear
    Non
    13-425
    Discontinued
    Supplier Number: -
  • Vials
    8.00 ml
    17×60 mm
    Clear
    Non
    15-425
    Discontinued
    Supplier Number: -
  • Vials
    12.00 ml
    19×65 mm
    Clear
    Non
    15-425
    Discontinued
    Supplier Number: -
  • Vials
    16.00 ml
    21×70 mm
    Clear
    Non
    18-400
    Discontinued
    Supplier Number: -
  • Vials
    22.00 ml
    23×85 mm
    Clear
    Non
    20-400
    Discontinued
    Supplier Number: -
  • Vials
    40.00 ml
    28×95 mm
    Clear
    Non
    24-400
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $252.53
    $252.53
    Pack of 100
       $252.53
    Supplier Number: B7999-6
  • Vials
    2.00 ml
    12×32 mm
    Amber
    Non
    8-425
    Discontinued
    Supplier Number: -
  • Vials
    8.00 ml
    17×60 mm
    Amber
    Non
    15-425
    Discontinued
    Supplier Number: B7999-3A
  • Vials
    12.00 ml
    19×65 mm
    Amber
    Non
    15-425
    Discontinued
    Supplier Number: -
  • Vials
    16.00 ml
    21×70 mm
    Amber
    Non
    18-400
    Discontinued
    Supplier Number: -
  • Vials
    40.00 ml
    28×95 mm
    Amber
    Non
    24-400
    Discontinued
    Supplier Number: -
  • Vials
    4.00 ml
    15×45 mm
    Amber
    Non
    13-425
    Discontinued
    Supplier Number: -
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • White PTFE/clear silicone septa, 5-425, thickness 2.6 mm
    White PTFE/clear silicone septa, 5-425, thickness 2.6 mm
    66030-760
    Log in pour la disponibilité
    Pack of 100 - $148.35
Informations sur le produit
  • Catalog #
    Material
    Cap Color
    Availability
    Price
  • Screw Caps, 18-400, PTFE/PE Foam Liner
    Screw Caps, 18-400, PTFE/PE Foam Liner
    66030-775
    PP
    White
    Log in pour la disponibilité
    Pack of 100 - $214.07
Informations sur le produit
  • Catalog #
    Material
    Cap Color
    Availability
    Price
  • Screw Caps, 20-400, PTFE/PE Foam Liner
    Screw Caps, 20-400, PTFE/PE Foam Liner
    66030-779
    PP
    White
    Log in pour la disponibilité
    Pack of 100 - $245.57