SP Bel-Art Poxygrid® Half-Size Test Tube Racks, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Excellent space-savers for use in water baths, laminar flow hoods, fume hoods or any place where space is limited.
- Compact racks
- Epoxy-coated steel wire construction
- Chemical and corrosion-resistant
- Unaffected by organic solvents, salts, weak acids, and weak alkalis
- Autoclavable
These compact racks have square holes, three decks, slotted mesh bottom deck, and reinforced edges.
Frequently Bought Together