Accessories for UVP ChemStudio SA2 Imager with BioLite MultiSpectral Light Source Promotion, Analytik Jena
Catalogue # 14220-782
Fournisseur: Analytik Jena US
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:DigiDoc-It Drawer, 5.8" x 19" x 13.2" (H x W x D)
- Cat. no.:14220-782
- Supplier No.:980068-01