SP Bel-Art Poxygrid® Pasteur Pipette Can Rack, Bel-Art Products, a part of SP
Catalog # 47729-700
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:4-Place Pasteur Pipette Rack
- Dimensions:19L x 14W x 19.4H cm (7½ x 5½ x 7⅝")
- Cat. no.:47729-700
- Supplier No.:189420000
Specifications
About this item
Designed for the safe, convenient storage of Pasteur Pipettes or transfer pipettes.
- Safe and convenient storage
- Durable
- Epoxy-coated steel wire construction
- Chemical- and corrosion-resistant
- Autoclavable
Individual compartments store up to four pipette sterilization cans, each with dimensions of 5.7 x 5.7 cm (2¼ x 2¼"). The durable rack sits flat on the benchtop and supports pipettes at an angle to allow for easy selection and use.
Epoxy-coated steel wire is chemical and corrosion resistant and steam autoclavable at 121°C (250°F). pipette cans not included.