Order Entry
Canada
ContactUsLinkComponent
Somso® Artificial Base of Skull with Arteries
  470106-726
undefined
Somso® Artificial Base of Skull with Arteries
  470106-726
 :  QS 65/6

 

  • Description:
    Base of skull arteries
  • Height:
    27 cm
  • Length:
    20 cm
  • Cat. no.:
    470106-726
  • Width:
    18 cm
  • Supplier No.:
    QS 65/6

 

 

Somso® premium quality models.

  • 5 year warranty

Natural casting in SOMSO-Plast®, consisting of green base of skull, mandible and cervical vertebrae with nerves. Representation of cervical arteries with internal passage through the green base of skull with emphasis on basilar artery with complete circle of Willisi. In one piece. Movable on stand with green base.

 : Items on this page are not stocked and may have an extended time for delivery.