Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Somso® Larynx

Larynx, 16 cm, 14 cm, 24.5 cm
  470231-960
 :  GS 7
Somso® Larynx
  470231-960
Somso® Larynx
Somso® Larynx

Larynx, 16 cm, 14 cm, 24.5 cm

 
$1,800.00
   
Each
 

Somso® premium quality models.

  • 5 year warranty

Enlarged approx. 2 times, in SOMSO-Plast®. Median section (2 parts). Removable parts: the right thyroid cartilage, the cricothyroid muscle and the thyrohyoid muscle. Inner and outer muscles of the larynx, the relief of mucous membrane, network of arteries and nerves and the cartilaginous skeleton can be demonstrated. Separates into 5 parts. On a stand with green base.

 : Items on this page are not stocked and may have an extended time for delivery.

 
  • Désignation:
    Larynx
  • Hauteur:
    24.5 cm
  • Longueur:
    14 cm
  • Width:
    16 cm