Order Entry
Canada
ContactUsLinkComponent
Somso® Helmet Orchid Flower
Somso® Helmet Orchid Flower
Catalog # 470312-422
Supplier:  MARCUS SOMMER
undefined
Somso® Helmet Orchid Flower
Catalog # 470312-422
Supplier:  MARCUS SOMMER
Supplier Number:  BOS14/10

Specifications

  • Description:
    Helmet orchid flower
  • Height:
    26 cm
  • Length:
    32 cm
  • Cat. no.:
    470312-422
  • Width:
    19 cm

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Orchis militaris, enlarged 13 times, in SOMSO-Plast®. After Prof. Dr. W. Weber. The model depicts the complicated structure of an orchid flower. Separable into 5 parts. On a stand with explanatory note on the green base.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.