Order Entry
Canada
ContactUsLinkComponent
Somso® Fruit of the Cacao
Catalog # 470312-448
Supplier:  MARCUS SOMMER
undefined
Somso® Fruit of the Cacao
Catalog # 470312-448
Supplier:  MARCUS SOMMER
Supplier Number:  BOS15/33

Specifications

  • Description:
    Fruit of the cacao
  • Height:
    30 cm
  • Length:
    17.5 cm
  • Cat. no.:
    470312-448
  • Width:
    17.5 cm

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Theobroma cacao, natural size, made of SOMSO-Plast®. According to Prof. Dr. W. Weber. Separable into 7 parts.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.