Somso® Model of the Heart with Bypass Vessels (Aortic Coronary Venous Bypass)
Catalog # 470313-116
Supplier: MARCUS SOMMER
Specifications
- Description:Heart with bypass vessels 2 Pt
- Height:28 cm
- Length:16.5 cm
- Cat. no.:470313-116
- Width:14 cm
Specifications
About this item
Somso® premium quality models.
- 5 year warranty
Natural size, in SOMSO-Plast®. Developed in co-operation with Prof. Dr. Meisner. The model shows one venous bypass leading to the right coronary artery as well as the descending anterior interventricular ramus (anterior wall) and the circumflex ramus of the left coronary artery. Separates into 2 parts. On a stand and green base.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.