ContactUsLinkComponent
Somso® Root Tip of A Monocotyledonous Plant In Longitudinal and Cross Section
Somso® Root Tip of A Monocotyledonous Plant In Longitudinal and Cross Section
  470312-540
undefined
Somso® Root Tip of A Monocotyledonous Plant In Longitudinal and Cross Section
  470312-540
 :  BOS20/2
  • Description:
    Monocot root CS LS
  • Height:
    37 cm
  • Length:
    18 cm
  • Cat. no.:
    470312-540
  • Width:
    18 cm
Somso® premium quality models.
  • 5 year warranty
Barley, Hordeum vulgare, enlarged approx. 200 times, in SOMSO-Plast®. After Prof. Dr. W. Weber. The root cap encloses the apical meristem. The tissues of the root body are formed by various initials: rhizodermis, root bark and central cylinder. In one piece, on a green base, with description.
 : Items on this page are not stocked and may have an extended time for delivery.