Order Entry
Canada
ContactUsLinkComponent
Somso® Buttercup Flower
Catalogue # 470312-438
Fournisseur:  MARCUS SOMMER
undefined
Somso® Buttercup Flower
Catalogue # 470312-438
Fournisseur:  MARCUS SOMMER
Supplier Number:  BOS15/20-A

Spécifications

  • Description:
    Buttercup flower
  • Height:
    34 cm
  • Length:
    26 cm
  • Cat. no.:
    470312-438
  • Width:
    26 cm

Spécifications

À propos de cet article

Somso® premium quality models.

  • 5 year warranty

Tall buttercup, Ranunculus acer, enlarged approx. 10 times, in SOMSO-Plast®. After Prof. Dr. W. Weber. In one piece. On a stand with green base and explanatory note.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.