Order Entry
Canada
ContactUsLinkComponent
 
Somso® Dandelion Blossom Fruit
  470312-536
undefined
Somso® Dandelion Blossom Fruit
  470312-536
 :  BOS15/19

 

  • Description:
    Dandelion blossom fruit
  • Height:
    36 cm
  • Length:
    18 cm
  • Cat. no.:
    470312-536
  • Width:
    33 cm

 

 

Somso® premium quality models.

  • 5 year warranty

Taraxacum officinale, enlarged approx. 8 times, made in SOMSO-Plast®. After Prof. Dr. W. Weber. The inflorescence is longitudinally sectioned. The individual flower and fruit can be removed from the green base. With explanatory note.

 : Items on this page are not stocked and may have an extended time for delivery.