Pliers Decappers, Wheaton | Avantor
Pliers Decappers, Wheaton
Pliers Decappers, Wheaton
Certificates
Pliers Decappers, Wheaton
Pliers Decappers, Wheaton
2 Options Available Below
Product Details & Documents
Pliers decappers are an economical and easy way to remove seals.
To remove a seal, simply secure the seal between the jaws, squeeze the handles and twist the seal. Constructed of steel with red plastic-coated handles.
Product Family Options
Informations sur le produit
  • Seal Size
    Availability
    Price
  • 13 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $221.35
    $221.35
    Prix promotionnel pour les nouveaux clients
    Case of 1
       $221.35
    Supplier Number: 224372
  • 20 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $229.90
    $229.90
    Prix promotionnel pour les nouveaux clients
    Each
       $229.90
    Supplier Number: 224373