Eisco® Model Lymph Node
Lymph nodes model
Catalog # 470356-230
Supplier: Wards
Fournisseur #: AM0306
Jump to:
Product Details & Documents
An easy to understand lymph node.
- Mounted on base
- Includes key card
- Enlarged size
Model shows incoming lymph vessel, germinal centre, sinus, outgoing lymph vessel, capsule, Trabecula, incoming blood supply artery, and vein for outgoing blood. Lymph nodes vary in diameter from 1 to 25 mm, although they can swell during infection or illness. They are covered in a fibrous capsule, they contain sinuses, where many scavenging white blood cells, called macrophages, ingest bacteria as well as other foreign matter and debris.
Spécifications
- Désignation:Lymph nodes model