Data-Trend Multipurpose Temperature Chart Recorders, Dickson® | Avantor
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
Supplier:   Dickson
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
Supplier:  Dickson
Certificates
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
7 Options Available Below
Product Details & Documents
Adaptable to a wide range of applications, these recorders feature a 15.2cm (6") readable chart, AC power with battery backup, user-selectable recording times and temperature ranges, and a remote probe
Increased shielding eliminates electronic noise interference and allows smoother pen movement.
Ordering information: Recorders include red pen, AC adapter, 9V battery, and 1.2m (4') bead wire probe. Charts must be purchased separately.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 97006-874
  • 97006-876
  • 97006-878
  • 61280-572
  • 61280-569
  • 61280-554
  • 61280-551
  • 61280-571
  • 61280-565
  • 61280-553
  • 61280-549
  • 61280-561
  • Temperature accuracy:
  • ±0.5% of chart range, ±1°C (±1.8°F) recorder only
  • ±0.5% of chart range, ±1°C (±1.8°F) recorder only
  • ±0.5% of chart range, ±1°C (±1.8°F) recorder only
  • Recording time:
  • 24-hour or 7-day (user-selectable)
  • 24-hour or 7-day (user-selectable)
  • 24-hour or 7-day (user-selectable)
  • Dimensions:
  • 19.2H x 19.2W x 8.3D cm (79/16 x 79/16 x 31/4")
  • 19.2H x 19.2W x 8.3D cm (79/16 x 79/16 x 31/4")
  • 19.2H x 19.2W x 8.3D cm (79/16 x 79/16 x 31/4")
  • Weight:
  • 980 g (35 oz.)
  • 980 g (35 oz.)
  • 980 g (35 oz.)
Spécifications
Product Family Options
Charts
Informations sur le produit
  • Désignation
    Plage de température
    Availability
    Price
  • 7-Day Chart
    –50...50 °C/°F
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $193.63
    $193.63
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $162.04
    $162.04
    Each
       $193.63
    Pack of 60
       $162.04
    Supplier Number: C651
    Spécifications:
    More
    Specifications
    Cat. No.
    61280-572
    Temperature accuracy
    Recording time
    Dimensions
    Weight
  • 7-Day Chart
    0...50 °C/°F or –50...0 °C/°F
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $196.06
    $196.06
    Pack of 60
       $196.06
    Supplier Number: C653
    Spécifications:
    More
    Specifications
    Cat. No.
    61280-569
    Temperature accuracy
    Recording time
    Dimensions
    Weight
  • 7-Day Chart
    0...250 °C/°F
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $192.29
    $192.29
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $160.95
    $160.95
    Each
       $192.29
    Pack of 60
       $160.95
    Supplier Number: C659
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    61280-554
    Temperature accuracy
    Recording time
    Dimensions
    Weight
    Small Business Enterprise
  • 7-Day Chart
    0...500 °C/°F
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $196.06
    $196.06
    Pack of 60
       $196.06
    Supplier Number: C661
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    61280-551
    Temperature accuracy
    Recording time
    Dimensions
    Weight
    Small Business Enterprise
  • 24-Hour Chart
    –50...50 °C/°F
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $196.06
    $196.06
    Pack of 60
       $196.06
    Supplier Number: C652
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    61280-571
    Temperature accuracy
    Recording time
    Dimensions
    Weight
    Small Business Enterprise
  • 24-Hour Chart
    0...50 °C/°F or –50...0 °C/°F
    Discontinued
    Supplier Number: -
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    61280-565
    Temperature accuracy
    Recording time
    Dimensions
    Weight
    Small Business Enterprise
  • 24-Hour Chart
    0...250 °C/°F
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $192.29
    $192.29
    Pack of 60
       $192.29
    Supplier Number: C660
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    61280-553
    Temperature accuracy
    Recording time
    Dimensions
    Weight
    Small Business Enterprise
  • 24-Hour Chart
    0...500 °C/°F
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $193.63
    $193.63
    Pack of 60
       $193.63
    Supplier Number: C662
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    61280-549
    Temperature accuracy
    Recording time
    Dimensions
    Weight
    Small Business Enterprise
  • 24-Hour Chart
    50...100 °C/°F
    Discontinued
    Supplier Number: -
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    61280-561
    Temperature accuracy
    Recording time
    Dimensions
    Weight
    Small Business Enterprise
Charts
Informations sur le produit
  • Catalog #
    Plage de température
    Désignation
    Availability
    Price
  • 32...100 °F
    24-Hour Chart
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $192.29
    $192.29
    Pack of 60
       $192.29
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Red Pens
    Red Pens
    61280-132
    Log in pour la disponibilité
    Pack of 6 - $244.14