VWR® Screw-Thread Vials, 4 ml
VWR® Screw-Thread Vials, 4 ml
VWR® Screw-Thread Vials, 4 ml
Supplier:  Thermo Scientific
Certificates
VWR® Screw-Thread Vials, 4 ml
VWR® Screw-Thread Vials, 4 ml
1 Options Available Below
Product Details & Documents
Designed for use with Waters® WISP® (48-position) and Shimadzu™ autosamplers.
  • 13-425 thread finish
  • ID vials feature write-on patch with graduation for convenient sample identification
  • Superior quality borosilicate glass
Caps have a central hole measuring 8.5 mm and a septa, ND13.
Product Family Options
Vials
Product Information
  • Désignation
    Capacité
    Diameter ×H
    Color
    Avec bouchon
    Taille du bouchon
    Cap Material
    Cap Color
    Septum
    Availability
    Price
  • Screw-Thread Vials
    4.00 ml
    15×45 mm
    Amber
    Non
    Discontinued
  • Screw-Thread Vials
    4.00 ml
    15×45 mm
    Clear
    Non
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 200
    Pack of 200 - $166.48
    $166.48
    Log in for availability
    Case of 1000
    Case of 1000 - $648.88
    $648.88
    New Customer Promotional Price
    Pack of 200
       $166.48
    Case of 1000
       $648.88
  • Screw-Thread Vials with Marking Spot
    4.00 ml
    15×45 mm
    Amber
    Non
    Discontinued
  • Screw-Thread Vials with Marking Spot
    4.00 ml
    15×45 mm
    Clear
    Non
    Discontinued