Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
 :   Dickson
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
 :  Dickson
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
Adaptable to a wide range of applications, these recorders feature a 15.2cm (6") readable chart, AC power with battery backup, and user-selectable recording times and temperature ranges
Increased shielding eliminates electronic noise interference and allows smoother pen movement.
Ordering information: Recorders include one red pen, one blue pen, AC adapter, and 9V battery. Charts must be purchased separately.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
  • Catalog No:
  • 97006-886
  • 61280-319
  • 61280-321
  • 61280-323
  • 61280-557
  • 61280-559
  • 61280-317
  • Temperature accuracy:
  • ±0.5% of chart range, ±1°C (±1.8°F) recorder only
  • Humidity range:
  • 0 to 95% RH
  • Humidity accuracy:
  • ±2% from 10 to 60%, ±3% from 60 to 95%
  • Recording time:
  • 24-hour or 7-day (user-selectable)
  • Dimensions:
  • 19.2H x 19.2W x 8.3D cm (79/16 x 79/16 x 31/4")
  • Weight:
  • 980 g (35 oz.)
Product Family Options
Charts
  • Désignation
    Plage de température
  • 24-Hour Chart
    0...50 °C
     : -
    Small Business Enterprise
    Specifications
    Cat. No.
    61280-319
    Temperature accuracy
    Humidity range
    Humidity accuracy
    Recording time
    Dimensions
    Weight
    Small Business Enterprise
  • 7-Day Chart
    0...185 °F/°C
    Pack of 60
    Pack of 60 - $192.29
    $192.29
    Pack of 60
       $192.29
     : C665
    Small Business Enterprise
    Specifications
    Cat. No.
    61280-321
    Temperature accuracy
    Humidity range
    Humidity accuracy
    Recording time
    Dimensions
    Weight
    Small Business Enterprise
  • 24-Hour Chart
    0...185 °F/°C
    Pack of 60
    Pack of 60 - $192.29
    $192.29
    Pack of 60
       $192.29
     : C667
    Small Business Enterprise
    Specifications
    Cat. No.
    61280-323
    Temperature accuracy
    Humidity range
    Humidity accuracy
    Recording time
    Dimensions
    Weight
    Small Business Enterprise
  • 24-Hour Chart
    32...100 °F
    Pack of 60
    Pack of 60 - $192.29
    $192.29
    Pack of 60
       $192.29
     : C658
    Small Business Enterprise
    Specifications
    Cat. No.
    61280-557
    Temperature accuracy
    Humidity range
    Humidity accuracy
    Recording time
    Dimensions
    Weight
    Small Business Enterprise
  • 7-Day Chart
    0...100 °F/°C
    Pack of 60
    Pack of 60 - $193.63
    $193.63
    Pack of 60
       $193.63
     : C657
    Specifications
    Cat. No.
    61280-559
    Temperature accuracy
    Humidity range
    Humidity accuracy
    Recording time
    Dimensions
    Weight
  • 7-Day Chart
    0...50 °C
    Pack of 60
    Pack of 60 - $242.32
    $242.32
    Pack of 60
       $242.32
     : C663
    Specifications
    Cat. No.
    61280-317
    Temperature accuracy
    Humidity range
    Humidity accuracy
    Recording time
    Dimensions
    Weight
Accessories
  •  
    Description
  • 13450-754
    NIST Traceable Calibration 1-Point Service
    Each - $291.26
  •  
    Description
  • 13450-756
    NIST & A2LA Traceable Calibration 3-Point Service
    Each - $848.52
  •  
    Description
  • 61280-134
    Pens, Three Red and Three Blue
    Pack of 6 - $244.14