Shell Vials
Shell Vials
Supplier:   Thermo Scientific
Shell Vials
Supplier:  Thermo Scientific
Certificates
Shell Vials
4 Options Available Below
Product Details & Documents
The 1 ml, 8×35 mm vials are designed for use in Alcott model 728 autosamplers. The 1 ml, 8×40 mm vials are specifically designed for Waters® 96-position trays. The 2 ml vial will work on any standard size 2 ml tray. The 4 ml vial is compatible with Waters® 48-position trays. Shell vials are manufactured from borosilicate glass unless otherwise specified.
  • Molded sealing ring secures sample containment
SepCap™ closures are designed for easy syringe needle penetration and easy push-in/pull-out from vial.
Conical-style Polyspring® inserts are available in borosilicate glass or polypropylene.
Certifications: Certified vial kits are tested and certified to meet or exceed up to 17 critical performance parameters; certificate enclosed with kit.
Delivery information: Vials are supplied with polyethylene push-in caps or polypropylene SepCap™ closures. Convenience kits include vials and caps with pre-inserted septa.
Product Family Options
Vials
Informations sur le produit
  • Désignation
    Capacité
    Bottom Style
    Diameter ×H
    Matière
    Color
    Avec bouchon
    Cap Type
    Cap Color
    Availability
    Price
  • Vials
    0.90 ml
    Tapered bottom
    8×40 mm
    PP
    Clear
    Oui
    SepCap™
    Natural
    Discontinued
    Supplier Number: C4015-94
  • Discontinued
    Supplier Number: -
  • Vials
    1.00 ml
    Flat
    8×32 mm
    Glass
    Clear
    Non
    Discontinued
    Supplier Number: 1-NWV-I
  • Vials
    1.00 ml
    Flat bottom
    8×40 mm
    Glass
    Clear
    Oui
    SepCap™
    Natural
    Discontinued
    Supplier Number: -
  • Vials
    1.00 ml
    Flat bottom
    8×35 mm
    Glass
    Clear
    Oui
    SepCap™
    Natural
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1,000
    Pack of 1,000 - $1,229.58
    $1,229.58
    Prix promotionnel pour les nouveaux clients
    Pack of 1,000
       $1,229.58
    Supplier Number: C4008-50
  • Vials
    1.00 ml
    Flat bottom
    8×40 mm
    Glass
    Amber
    Oui
    SepCap™
    Natural
    Discontinued
    Supplier Number: -
  • Vials
    1.00 ml
    Flat bottom
    8×40 mm
    PP
    Clear
    Oui
    SepCap™
    Natural
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 250
    Pack of 250 - $163.36
    $163.36
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - $653.45
    $653.45
    Prix promotionnel pour les nouveaux clients
    Pack of 250
       $163.36
    Case of 1000
       $653.45
    Supplier Number: C4015-95P
  • Vials
    2.00 ml
    Flat bottom
    12×32 mm
    Glass
    Clear
    Oui
    SepCap™
    Natural
    Discontinued
    Supplier Number: -
  • Vials, conical bottom
    2.00 ml
    Conical bottom
    12×32 mm
    PP
    Clear
    Oui
    SepCap™
    Natural
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $931.23
    $931.23
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $931.23
    Supplier Number: C4011-77P
  • Vials
    4.00 ml
    Flat bottom
    15×45 mm
    Glass
    Clear
    Oui
    SepCap™
    Natural
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $81.04
    $81.04
    Prix promotionnel pour les nouveaux clients
    Pack of 100
       $81.04
    Supplier Number: C4015-48