700 System Premium Receiver Head Assemblies, Techcon
Supplier: Techcon Systems
Jump to:
À propos de cet article
Head assemblies ensure accident-proof dispensing and provide a secure and reliable link between the dispensing barrel and the controller
Complete dispensing control. Compatible with all industry-standard dispensing barrels. Designed to exact industry standards.
7 Options Available Below
Product Details & Documents
Head assemblies ensure accident-proof dispensing and provide a secure and reliable link between the dispensing barrel and the controller
Complete dispensing control. Compatible with all industry-standard dispensing barrels. Designed to exact industry standards.
Product Family Options
Assembly for 3 cc Barrel
Informations sur le produit
Assembly for 5 cc Barrel
Informations sur le produit
Assembly for 10 cc Barrel
Informations sur le produit
- Longueur TubesAvailabilityPrice
- Supplier Number: 71006RHB
Assembly for 30 cc/55 cc Barrel