700 System Premium Pistons, Techcon
À propos de cet article
Pistons are available in white wiper and red straight wall styles
The low friction wiper piston can be used with all materials and features a full lip seal. The air-free loading design of the straight wall piston allows air to bleed while retaining product security. Made of polyethylene. UV blocking. Dispense high-viscosity materials of 55,000cP minimum.
8 Options Available Below
Product Details & Documents
Pistons are available in white wiper and red straight wall styles
The low friction wiper piston can be used with all materials and features a full lip seal. The air-free loading design of the straight wall piston allows air to bleed while retaining product security. Made of polyethylene. UV blocking. Dispense high-viscosity materials of 55,000cP minimum.
Product Family Options
White Wiper
Informations sur le produit
- CapacitéAvailabilityPrice
- Supplier Number: 7100009WPK
- Supplier Number: 7030009WPK
- Supplier Number: 7300009WPK
- Supplier Number: 7050009WPK
Red Straight Wall
Informations sur le produit
- CapacitéAvailabilityPrice
- Supplier Number: 7030006RPK
- Supplier Number: 7050006RPK
- Supplier Number: 7100006RPK
- Supplier Number: 7300006RPK