SE Series Dispensing Tips, Techcon | Avantor
SE Series Dispensing Tips, Techcon
Supplier:   Techcon Systems
SE Series Dispensing Tips, Techcon
Supplier:  Techcon Systems
Certificates
SE Series Dispensing Tips, Techcon
SE Series Dispensing Tips, Techcon
2 Options Available Below
Product Details & Documents
Tips have a polypropylene hub and stainless steel cannula
Tips are 1.27cm (1/2") long. Colors indicate gauge size.
Product Family Options
Informations sur le produit
  • Color
    Jauge
    Availability
    Price
  • Green
    18
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $73.64
    $73.64
    Pack of 50
       $73.64
    Supplier Number: SE418050PK
  • Red
    25
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $73.64
    $73.64
    Pack of 50
       $73.64
    Supplier Number: SE425050PK