PYREX® Cylindrical Jars, Corning
PYREX® Cylindrical Jars, Corning
Supplier:   Corning
PYREX® Cylindrical Jars, Corning
Supplier:  Corning
Certificates
PYREX® Cylindrical Jars, Corning
PYREX® Cylindrical Jars, Corning
4 Options Available Below
Product Details & Documents
These PYREX® jars are mold-blown with substantial walls for mechanical strength.
  • Open end is ground flat
These jars can also be used as animal jars.
Caution: Do not use with heat, vacuum, or pressure applications.
Product Family Options
Product Information
  • Désignation
    Capacité
    Ø ext.
    Hauteur
    Availability
    Price
  • Plain Jars
    9.5 L (2.5 gal.)
    25.7 cm (101/8")
    25.4 cm (10")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $1,100.00
    $1,100.00
    Case of 1
       $1,100.00
    Supplier Number: 6942-9L
  • Plain Jars
    17 L (4.5 gal.)
    30.5 cm (12")
    30.5 cm (12")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $2,110.00
    $2,110.00
    Case of 1
       $2,110.00
    Supplier Number: 6942-17L
  • Plain Jars
    26.5 L (7 gal.)
    30.5 cm (12")
    45.7 cm (18")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $1,730.00
    $1,730.00
    Case of 1
       $1,730.00
    Supplier Number: 6942-27L
  • Plain Jars
    32 L (8.4 gal.)
    40.6 cm (16")
    30.5 cm (12")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $2,770.00
    $2,770.00
    Case of 1
       $2,770.00
    Supplier Number: 6942-32L