PYREX® Rectangular Chromatography Jars, Corning | Avantor
PYREX® Rectangular Chromatography Jars, Corning
Supplier:   Corning
PYREX® Rectangular Chromatography Jars, Corning
Supplier:  Corning
Certificates
PYREX® Rectangular Chromatography Jars, Corning
PYREX® Rectangular Chromatography Jars, Corning
2 Options Available Below
Product Details & Documents
Jars are ground to the close tolerance needed for a tight cover fit.
  • These rectangular chromatography jars are made of Pyrex® borosilicate glass
Edges are ground flat within 0,25mm (0,01").
 : Covers are not supplied.
 : Not recommended for use with direct heat, pressure, or vacuum applications.
Product Family Options
Informations sur le produit
  • Capacité
    W×D×H
    Availability
    Price
  • 3.8 L (1 gal.)
    13.7 x 16.2 x 26.7 cm (53/8 x 63/8 x 101/2")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $3,280.00
    $3,280.00
    Case of 6
       $3,280.00
    Supplier Number: 6944-4L
  • 11.4 L (3 gal.)
    23.8 x 18.1 x 32.4 cm (93/8 x 71/8 x 123/4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $1,350.00
    $1,350.00
    Case of 1
       $1,350.00
    Supplier Number: 6944-11L