PYREX® Cylindrical Chromatography Jars, Corning | Avantor
PYREX® Cylindrical Chromatography Jars, Corning
Supplier:   Corning
PYREX® Cylindrical Chromatography Jars, Corning
Supplier:  Corning
Certificates
PYREX® Cylindrical Chromatography Jars, Corning
PYREX® Cylindrical Chromatography Jars, Corning
3 Options Available Below
Product Details & Documents
These general purpose cylindrical chromatography jar with tops ground to close tolerances to insure a tight cover fit. Covers are not supplied.
These cylindrical chromatography jars are made of Pyrex® borosilicate glass.
The jar edge is flat within 0.102 mm (0.004") on 6.6 L (1.7 gal.) jar, all others are flat within 0.254 mm (0.01").
Caution: Note: Do not use with heat, vacuum, or pressure applications.
Product Family Options
Informations sur le produit
  • Capacité
    Ø ext.
    Hauteur
    Availability
    Price
  • 6.6 L (1.7 gal.)
    15.2 cm (6")
    45.7 cm (18")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $761.00
    $761.00
    Case of 1
       $761.00
    Supplier Number: 6945-6L
  • 13.2 L (3.5 gal.)
    22.2 cm (83/4")
    45.7 cm (18")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $4,620.00
    $4,620.00
    Case of 4
       $4,620.00
    Supplier Number: 6945-13L
  • 36 L (9.5 gal.)
    30.5 cm (12")
    61 cm (24")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $3,220.00
    $3,220.00
    Case of 1
       $3,220.00
    Supplier Number: 6945-36L