PYREX® Animal Jars, Corning
PYREX® Animal Jars, Corning
Supplier:   Corning
PYREX® Animal Jars, Corning
Supplier:  Corning
Certificates
PYREX® Animal Jars, Corning
PYREX® Animal Jars, Corning
2 Options Available Below
Product Details & Documents
PYREX® glass.
Ideal for use as animal jars or for other purposes where resistance to hot air and steam sterilization is of importance. The open ends are finished with a fire polished bead to increase mechanical strength.
Caution: Note: Do not use with heat, vacuum, or pressure applications.
Product Family Options
Product Information
  • Matière
    Capacité
    Ø ext.
    Hauteur
    Épaisseur
    Availability
    Price
  • PYREX® borosilicate glass
    2.8 L (94.7 oz.)
    15.2 cm (6")
    20.3 cm (8")
    0.48 cm (³⁄₁₆")
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $489.00
    $489.00
    Case of 1
       $489.00
    Supplier Number: 6941-3L
  • PYREX® borosilicate glass
    4.7 L (1.2 gal.)
    21 cm (81/4")
    20.3 cm (8")
    0.48 cm (³⁄₁₆")
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $2,790.00
    $2,790.00
    Case of 6
       $2,790.00
    Supplier Number: 6941-5L