PYREX® Bell Jars, Corning
PYREX® Bell Jars, Corning
 :   Corning
PYREX® Bell Jars, Corning
 :  Corning
PYREX® Bell Jars, Corning
PYREX® Bell Jars, Corning
These specially blown, sturdy, round-bottom jars have a fine ground finish at the open end. Flat, ground surface has a contact area equal to the wall thickness at the open end.
Caution: Do not use with heat, pressure or vacuum applications.
Product Family Options
  • Hauteur
    Ø ext.
  • 305 mm (12")
    318 mm (121/2")
    Case of 1
    Case of 1 - $3,130.00
    $3,130.00
    Case of 1
       $3,130.00
     : 6886-12L
  • 457 mm (18")
    457 mm (18")
    Case of 1
    Case of 1 - $6,190.00
    $6,190.00
    Case of 1
       $6,190.00
     : 6886-49L